3-(3-Methoxyphenyl)propionic acid ≥99%

Synonyms: 3-(3-Methoxyphenyl)propanoic acid

349763-5G 349763-25G
220059-928EA 42 USD
220059-928 220059-927
3-(3-Methoxyphenyl)propionic acid ≥99%
3-(3-Methoxyphenyl)propionic acid
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
MDL Number: MFCD00014027
CAS Number: 10516-71-9
EINECS: 234-049-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR