6-Fluoroindole

Warning

Synonyms: 6-Fluoro-1H-indole

349593-5G 349593-1G
220059-892EA 77.65 USD
220059-892 220059-891
6-Fluoroindole
6-Fluoroindole
Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 72…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00056933
CAS Number: 399-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR