Boron trichloride 1 M in heptane

348325-800ML 348325-100ML
220059-061EA 234 USD
220059-061 220059-550
Boron trichloride 1 M in heptane
Boron trichloride
Formula: BCl₃
MW: 117.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011313
CAS Number: 10294-34-5
EINECS: 233-658-4
UN: 3384
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR