2-Chloro-4-fluorobenzaldehyde ≥97%

Warning

348074-5G 348074-1G
220059-497EA 31.77 USD
220059-497 220059-496
2-Chloro-4-fluorobenzaldehyde ≥97%
2-Chloro-4-fluorobenzaldehyde
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 60 °C
MDL Number: MFCD00042527
CAS Number: 84194-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR