Dibenzothiophene

Synonyms: Dibenzo[b,d]thiophene

347833-5G 347833-1G
220058-607EA 105.22 USD
220058-607 220058-606
Dibenzothiophene
Dibenzothiophene
Formula: C₁₂H₈S
MW: 184.26 g/mol
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 95…98 °C
Density: 1.1295 g/cm³ (125 °C)
Flash Pt: 170 °C
MDL Number: MFCD00004969
CAS Number: 132-65-0
EINECS: 205-072-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR