Trifluoromethanesulfonic acid ≥99%, ReagentPlus®

Synonyms: Trifluoromethanesulphonic acid

347817-5G 347817-25G
220058-602EA 30.11 USD
220058-602 220058-601
Trifluoromethanesulfonic acid ≥99%, ReagentPlus®
Trifluoromethanesulphonic acid
Formula: CHF₃O₃S
MW: 150.08 g/mol
Boiling Pt: 162 °C (1013 hPa)
Melting Pt: 35 °C
Density: 1.69 g/cm³ (20 °C)
MDL Number: MFCD00007514
CAS Number: 1493-13-6
EINECS: 216-087-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR