Disulfiram

Warning

Synonyms: Tetraethylthiuram disulphide, 1,1',1'',1'''-{Disulphanediylbis[(thioxomethylene)-nitrilo]}tetraethane, Bis(diethylthiocarbamoyl) disulphide

86720-50G 86720-1KG 86720-250G
220098-699EA 66.45 USD
220098-699 220098-697 220098-698
Disulfiram
Disulphiram
Formula: C₁₀H₂₀N₂S₄
MW: 296.55 g/mol
Boiling Pt: 369 °C (1013 hPa)
Melting Pt: 70 °C
Density: 1.34 g/cm³ (20 °C)
MDL Number: MFCD00009048
CAS Number: 97-77-8
EINECS: 202-607-8
UN: 3077
ADR: 9,III
Merck Index: 13,03399

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR