(1S,2S)-(+)-1,2-Diaminocyclohexane ≥98%

Synonyms: (S,S)-DACH, (1S,2S)-Cyclohexane-1,2-diamine

346713-5G 346713-1G
220059-048EA 320.92 USD
220059-048 220059-047
(1S,2S)-(+)-1,2-Diaminocyclohexane ≥98%
(1S,2S)-(+)-1,2-Diaminocyclohexane
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: 40…43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00062986
CAS Number: 21436-03-3
UN: 3259
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR