2-Phenoxyaniline ≥99%

Synonyms: 1-Amino-2-phenoxybenzol

346683-500G 346683-25G
220059-043EA 324.15 USD
220059-043 220059-352
2-Phenoxyaniline ≥99%
2-Phenoxyaniline
Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
Boiling Pt: 170 °C (18 -rr)
Melting Pt: 46 - 48 °C
MDL Number: MFCD00035765
CAS Number: 2688-84-8
EINECS: 220-254-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR