4-Nitropyridine-N-oxide

Synonyms: 4-Nitropyridine N-oxide

346659-100G 346659-25G
220059-348EA 89.08 USD
220059-348 220059-349
4-Nitropyridine-N-oxide
4-Nitropyridine-N-oxide
Formula: C₅H₄N₂O₃
MW: 140.1 g/mol
Boiling Pt: 397 °C (1013 hPa)
Melting Pt: 161 °C
Density: 1.52 g/cm³ (20 °C)
MDL Number: MFCD00006206
CAS Number: 1124-33-0
EINECS: 214-395-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR