1,2-Naphthoquinone

Warning

346616-5G 346616-1G
220059-345EA 458.46 USD
220059-345 220059-344
1,2-Naphthoquinone
1,2-Naphthoquinone
Formula: C₁₀H₆O₂
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 145…146 °C
Density: 1.45 g/cm³ (20 °C)
MDL Number: MFCD00001698
CAS Number: 524-42-5
EINECS: 208-360-2
Merck Index: 12,06481

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR