Zirconyl nitrate hydrate, Technical Grade

Danger

Synonyms: Zirconium(IV) oxynitrate hydrate, Zirconium(IV) dinitrate oxide hydrate, Zirconium dinitrate oxide hydrate

346462-100G 346462-500G
220059-332EA 163.08 USD
220059-332 220059-333
Zirconyl nitrate hydrate, Technical Grade
Zirconyl nitrate hydrate
Formula: N₂O₇Zr·H₂O
MW: 249.25 g/mol
Density: 2.04 g/cm³ (20 °C)
MDL Number: MFCD00149896
CAS Number: 14985-18-3
EINECS: 237-529-3
UN: 1477
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR