Tetrabutylammonium chloride xhydrate

Warning

Synonyms: Tetrabutyl ammonium chloride xhydrate, Tetra-n-butylammonium chloride hydrate, Tetrabutylammonium chloride hydrate, TBAC

345857-5G 345857-100G 345857-25G
220059-262EA 28.38 USD
220059-262 220059-260 220059-261
Tetrabutylammonium chloride xhydrate
Tetrabutylammonium chloride xhydrate
Formula: C₁₆H₃₆ClN·xH₂O
MW: 277.92 g/mol (anhy)
MDL Number: MFCD00011635
CAS Number: 37451-68-6
EINECS: 214-195-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR