4-Fluoro-2-nitrophenol ≥99%

345059-25G 345059-5G
220059-127EA 32.31 USD
220059-127 220059-128
4-Fluoro-2-nitrophenol ≥99%
4-Fluoro-2-nitrophenol
Formula: C₆H₄FNO₃
Melting Pt: 74 °C
MDL Number: MFCD00042528
CAS Number: 394-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR