Triethylammonium dihydrogentrifluoride

Synonyms: Triethylamine trihydrofluoride, Triethylamine trishydrofluoride

344648-5G 344648-100G 344648-25G
220058-590EA 29.72 USD
220058-590 220058-588 220058-589
Triethylammonium dihydrogentrifluoride
Triethylammonium dihydrogentrifluoride
Formula: C₆H₁₈F₃N
MW: 161.21 g/mol
Density: 1.094 g/cm³ (20 °C)
MDL Number: MFCD00043294
CAS Number: 73602-61-6
EINECS: 277-550-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR