Lithium methoxide ≥98%

Danger

Synonyms: Lithium methanolate, Lithium methylate

344370-100G 344370-25G
220058-999EA 346.15 USD
220058-999 220059-000
Lithium methoxide ≥98%
Lithium methoxide
Formula: CH₃LiO
MW: 37.98 g/mol
Boiling Pt: 64.6 °C (1013 hPa)
Density: 0.865 g/cm³ (20 °C)
MDL Number: MFCD00036357
CAS Number: 865-34-9
EINECS: 212-737-7
UN: 3206
ADR: 4.2,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR