ω-Bromo-3-nitroacetophenone ≥97%

Synonyms: 3-Nitrophenacyl bromide, 2-Bromo-3'-nitroacetophenone, α-Bromo-3-nitroacetophenone, Bromomethyl m-nitrophenyl ketone

344214-5G 344214-25G
220058-879EA 73.23 USD
220058-879 220058-878
ω-Bromo-3-nitroacetophenone ≥97%
ω-Bromo-3-nitroacetophenone
Formula: C₈H₆BrNO₃
MW: 244.04 g/mol
Melting Pt: 90 - 94 °C
MDL Number: MFCD00024512
CAS Number: 2227-64-7
EINECS: 218-764-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR