Strontium ≥99%

343730-10G 343730-50G
220058-166EA 229.23 USD
220058-166 220058-167
Strontium ≥99%
Strontium
Formula: Sr
MW: 87.62 g/mol
Boiling Pt: 1151 °C (1013 hPa)
Melting Pt: 570 - 660 °C
Density: 2.38 g/cm³ (770 °C)
MDL Number: MFCD00134060
CAS Number: 7440-24-6
EINECS: 231-133-4
UN: 1393
ADR: 4.3,II
Merck Index: 12,08994

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR