(R)-(+)-Citronellal ≥90%, Technical Grade

Synonyms: (R)-(+)-3,7-Dimethyloct-6-enal

343641-5G 343641-1G
220058-156EA 599.85 USD
220058-156 220058-155
(R)-(+)-Citronellal ≥90%, Technical Grade
(R)-(+)-Citronellal
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
CAS Number: 2385-77-5
EINECS: 219-194-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR