1-Boc-piperazine

Synonyms: tert-Butyl 1-piperazinecarboxylate, tert-butyl piperazine-1-carboxylate

343536-25G 343536-1G 343536-5G
220058-833EA 314.46 USD
220058-833 220058-832 220058-834
1-Boc-piperazine
1-Boc-piperazine
Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
MDL Number: MFCD00075265
CAS Number: 57260-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR