Cobalt(II) oxide

Warning

Synonyms: Cobalt oxide

343153-100G 343153-10G
220058-814EA 1113.85 USD
220058-814 220058-815
Cobalt(II) oxide
Cobalt(II) oxide
Formula: CoO
MW: 74.93 g/mol
Melting Pt: 1933 °C
Density: 6.1 g/cm³ (20 °C)
MDL Number: MFCD00016031
CAS Number: 1307-96-6
EINECS: 215-154-6
UN: 2811
ADR: 6.1,III
Merck Index: 12,02507

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR