3-Fluoro-2-methylbenzaldehyde ≥98%

Synonyms: 3-Fluoro-o-tolualdehyde, 3-Fluoro-2-tolualdehyde, 3-Fluoro-o-tolylaldehyde

342521-25G 342521-1G 342521-5G
220058-561EA 284.62 USD
220058-561 220058-560 220058-562
3-Fluoro-2-methylbenzaldehyde ≥98%
3-Fluoro-2-methylbenzaldehyde
Formula: C₈H₇FO
MW: 138.14 g/mol
MDL Number: MFCD00075256
CAS Number: 147624-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR