Sodium phosphate ≥96%

Danger

Synonyms: tri-Sodium phosphate, Sodium phosphate tribasic, Trisodium phosphate

342483-2.5KG 342483-500G
220058-553EA 196.92 USD
220058-553 220058-555
Sodium phosphate ≥96%
Sodium phosphate
Formula: Na₃O₄P
MW: 163.94 g/mol
Melting Pt: 73.3…76.7 °C
Density: 2.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003510
CAS Number: 7601-54-9
EINECS: 231-509-8
UN: 3262
ADR: 8,III
Merck Index: 12,08808

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR