Copper(II) sulfide

Synonyms: Copper sulphide, Cupric sulphide, Copper(II)sulphide

342467-500G 342467-100G
220058-765EA 647.69 USD
220058-765 220058-764
Copper(II) sulfide
Copper(II) sulphide
Formula: CuS
MW: 95.61 g/mol
Density: 4.6 g/cm³ (20 °C)
MDL Number: MFCD00016066
CAS Number: 1317-40-4
EINECS: 215-271-2
Merck Index: 12,02724

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR