Copper (I) sulfide

Synonyms: Dicopper sulphide, Chacolcite, Copper(i)sulphide

342459-1KG 342459-250G
220058-761EA 1529.23 USD
220058-761 220058-762
Copper (I) sulfide
Copper (I) sulphide
Formula: Cu₂S
MW: 159.16 g/mol
Melting Pt: 1100 °C
Density: 5.783 g/cm³ (25 °C)
MDL Number: MFCD00016062
CAS Number: 22205-45-4
EINECS: 244-842-9
Merck Index: 13,02697

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR