2,7-Dibromofluorene

Warning

Synonyms: 2,7-Dibromo-9H-fluorene

342297-5G 342297-25G
220058-736EA 26.92 USD
220058-736 220058-735
2,7-Dibromofluorene
2,7-Dibromofluorene
Formula: C₁₃H₈Br₂
MW: 324.01 g/mol
MDL Number: MFCD00019048
CAS Number: 16433-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR