Triammonium dodecamolybdophosphate hydrate

Warning

Synonyms: Triammonium 12-molybdophosphate hydrate, Phosphomolybdic acid, ammonium salt hydrate, Ammonium phosphomolybdate hydrate

342165-50G 342165-250G
220058-711EA 173.85 USD
220058-711 220058-710
Triammonium dodecamolybdophosphate hydrate
Triammonium dodecamolybdophosphate hydrate
Formula: H₁₂Mo₁₂N₃O₄₀P·H₂O
MW: 1894.36 g/mol
MDL Number: MFCD00150423
CAS Number: 54723-94-3
EINECS: 234-714-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR