3-Fluoro-o-toluic acid ≥99%

342114-1G 342114-5G
220058-701EA 73.34 USD
220058-701 220058-702
3-Fluoro-o-toluic acid ≥99%
3-Fluoro-o-toluic acid
Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD00075246
CAS Number: 699-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR