Copper(II) acetate hydrate ≥98%

341746-500G 341746-100G 341746-2.5KG
220058-190EA 206.15 USD
220058-190 220058-665 220058-189
Copper(II) acetate hydrate ≥98%
Copper(II) acetate hydrate
Formula: C₄H₆CuO₄·H₂O
MW: 199.65 g/mol
Storage Temperature: Ambient
CAS Number: 66923-66-8
EINECS: 205-553-3
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR