Bismuth(III) iodide ≥99%

Danger

Synonyms: Bismuth triiodide

341010-100G 341010-25G
220058-492EA 251.31 USD
220058-492 220058-493
Bismuth(III) iodide ≥99%
Bismuth(III) iodide
Formula: BiI₃
MW: 589.69 g/mol
Melting Pt: 408 °C
Density: 5.78 g/cm³ (20 °C)
MDL Number: MFCD00010894
CAS Number: 7787-64-6
EINECS: 232-127-4
UN: 3260
ADR: 8,III
Merck Index: 12,01308

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR