Tetraethyl orthosilicate (TEOS)

Danger

Synonyms: Tetraethoxysilane

86578-250ML 86578-1L
220098-674EA 36.55 USD
220098-674 220098-673
Tetraethyl orthosilicate (TEOS)
Tetraethyl orthosilicate (TEOS)
Formula: Si(OC₂H₅)₄
MW: 208.33 g/mol
Boiling Pt: 168 °C (1013 hPa)
Melting Pt: –85.5 °C
Density: 0.933 g/cm³ (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00009062
CAS Number: 78-10-4
EINECS: 201-083-8
UN: 1292
ADR: 3,III
Merck Index: 13,03882

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR