4-Amino-5-chloro-2-methoxybenzoic acid ≥97%

Synonyms: Metoclopramide Impurity C, 4-Amino-5-chloro-o-anisic acid

340871-25G 340871-5G
220058-274EA 133.54 USD
220058-274 220058-275
4-Amino-5-chloro-2-methoxybenzoic acid ≥97%
4-Amino-5-chloro-2-methoxybenzoic acid
Formula: C₈H₈ClNO₃
MW: 201.61 g/mol
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 213 - 215 °C
MDL Number: MFCD00038794
CAS Number: 7206-70-4
EINECS: 230-582-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR