3-Methoxyanthranilic acid ≥98%

Warning

Synonyms: 2-Amino-m-anisic acid, 3-Methoxyanthranilic acid, o-Anisidine-6-carboxylic acid, 2-Amino-3-methoxybenzoic acid

340103-1G 340103-25G 340103-5G
220058-430EA 1136.92 USD
220058-430 220058-431 220058-432
3-Methoxyanthranilic acid ≥98%
3-Methoxyanthranilic acid
Formula: C₈H₉NO₃
MW: 167.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075178
CAS Number: 3177-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR