3,5-Dimethoxybenzylamine ≥98%

Synonyms: 3,5-Dimethoxybenzyl amine

339806-25G 339806-5G
220057-655EA 50.05 USD
220057-655 220057-656
3,5-Dimethoxybenzylamine ≥98%
3,5-Dimethoxybenzylamine
Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
MDL Number: MFCD00052813
CAS Number: 34967-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR