3,4-Dibutoxy-3-cyclobutene-1,2-dione ≥98%

Synonyms: Squaric acid dibutylester, Dibutyl squarate, 3,4-Dibutoxycyclobut-3-ene-1,2-dione, 1,2-Dibutoxy-1-cyclobutene-3,4-dione

339792-5G 339792-25G
220057-654EA 100.58 USD
220057-654 220057-653
3,4-Dibutoxy-3-cyclobutene-1,2-dione ≥98%
3,4-Dibutoxy-3-cyclobutene-1,2-dione
Formula: C₁₂H₁₈O₄
MW: 226.27 g/mol
Boiling Pt: 121 °C (1013 hPa)
Density: 0.96 g/cm³ (20 °C)
MDL Number: MFCD00037150
CAS Number: 2892-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR