Hydrobromic acid ≥99.99%

Danger

339245-100ML 339245-500ML
220058-319EA 117.38 USD
220058-319 220058-320
Hydrobromic acid ≥99.99%
Hydrobromic acid
Formula: HBr
Density: 1.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8,II

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR