Germanium(IV) ethoxide ≥99.95%

Synonyms: Germanium tetraethanolate, Germanic acid tetraethyl ester, Tetraethyl orthogermanate, Germanium tetraethoxide, Germanium (IV) ethanolate

339180-5G 339180-25G
220058-260EA 269.23 USD
220058-260 220058-259
Germanium(IV) ethoxide ≥99.95%
Germanium(IV) ethoxide
Formula: C₈H₂₀GeO₄
MW: 252.85 g/mol
Boiling Pt: 185…187 °C (1013 hPa)
Melting Pt: –72 °C
Density: 1.14 g/cm³ (20 °C)
MDL Number: MFCD00015121
CAS Number: 14165-55-0
EINECS: 238-007-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR