Diboron trioxide ≥99.98%

Danger

Synonyms: Boron oxide, Boric anhydride

339075-500G 339075-100G
220058-245EA 706.15 USD
220058-245 220058-244
Diboron trioxide ≥99.98%
Diboron trioxide
Formula: B₂O₃
MW: 69.62 g/mol
Boiling Pt: 1860 °C (1013 hPa)
Melting Pt: 450 °C
Density: 1.81 to 1.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011315
CAS Number: 1303-86-2
EINECS: 215-125-8
REACH: 01-2119486655-24
Merck Index: 13,01327
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52d3b9c48cafcf926185d4b331850a9d90d6e6b2&vid=52d3b9c48cafcf926185d4b331850a9d90d6e6b2&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR