Antimony(V) chloride ≥99.99%

Danger

Synonyms: Antimony pentachloride

338877-25G 338877-5G
220057-482EA 270.77 USD
220057-482 220057-930
Antimony(V) chloride ≥99.99%
Antimony(V) chloride
Formula: Cl₅Sb
MW: 299.02 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: 3 °C
Density: 2.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011213
CAS Number: 7647-18-9
EINECS: 231-601-8
UN: 1730
ADR: 8,II
Merck Index: 13,00701

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR