Ammonium fluoride

Danger

Synonyms: Fluoroammonium

338869-25G 338869-100G
220057-480EA 178.46 USD
220057-480 220057-479
Ammonium fluoride
Ammonium fluoride
Formula: NH₄F
MW: 37.04 g/mol
Melting Pt: 160 °C
Density: 1.0088 to 1.0096 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011423
CAS Number: 12125-01-8
EINECS: 235-185-9
UN: 2505
ADR: 6.1,III
Merck Index: 13,00524

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR