3-Phenoxyaniline ≥98%

Synonyms: 1-Amino-3-phenoxybenzol

338354-5G 338354-25G
220058-069EA 72.46 USD
220058-069 220058-068
3-Phenoxyaniline ≥98%
3-Phenoxyaniline
Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
Boiling Pt: 116 - 120 °C (0.3 -rr)
Melting Pt: 37 °C
Density: 1.1584 g/cm³ (25 °C)
MDL Number: MFCD00041891
CAS Number: 3586-12-7
EINECS: 222-715-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR