Ytterbium(III) chloride hexahydrate ≥99.9%

337927-10G 337927-50G
220058-027EA 90.92 USD
220058-027 220058-028
Ytterbium(III) chloride hexahydrate ≥99.9%
Ytterbium(III) chloride hexahydrate
Formula: Cl₃Yb·6H₂O
MW: 387.49 g/mol
Melting Pt: 154 °C
Density: 2.57 g/cm³ (20 °C)
MDL Number: MFCD00149885
CAS Number: 10035-01-5
EINECS: 233-800-5

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR