Copper(II)-2-ethylhexanoate

Synonyms: 2-Ethylhexanoic acid copper (II) salt, Copper(II) 2-ethylhexanoate, 2-Ethylcaproic acid copper (II) salt, Copper bis(2-ethylhexanoate)

337323-5G 337323-25G
220058-000EA 75.38 USD
220058-000 220057-999
Copper(II)-2-ethylhexanoate
Copper(II)-2-ethylhexanoate
Formula: C₁₆H₃₀CuO₄
MW: 349.96 g/mol
Melting Pt: 252 °C
MDL Number: MFCD00015695
CAS Number: 149-11-1
EINECS: 205-731-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR