4-Fluoro-m-cresol ≥98%

Warning

337250-25G 337250-5G
220057-989EA 193.85 USD
220057-989 220057-990
4-Fluoro-m-cresol ≥98%
4-Fluoro-m-cresol
Formula: C₇H₇FO
MW: 126.13 g/mol
MDL Number: MFCD00042296
CAS Number: 452-70-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR