2-Fluoro-p-toluidine ≥99%

Synonyms: 2-Fluoro-4-methylaniline, 4-Amino-3-fluorotoluene

337110-1G 337110-5G
220057-978EA 22.62 USD
220057-978 220057-979
2-Fluoro-p-toluidine ≥99%
2-Fluoro-p-toluidine
Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.1084 g/cm³ (18 °C)
MDL Number: MFCD00040975
CAS Number: 452-80-2
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR