4-(Trifluoromethoxy)aniline ≥98%

337056-25G 337056-5G
220057-959EA 84.75 USD
220057-959 220057-960
4-(Trifluoromethoxy)aniline ≥98%
4-(Trifluoromethoxy)aniline
Formula: C₇H₆F₃NO
MW: 177.13 g/mol
Boiling Pt: 73 °C (1013 hPa)
Density: 1.31 g/cm³ (20 °C)
MDL Number: MFCD00041314
CAS Number: 461-82-5
EINECS: 207-317-5
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR