Iridium(III) chloride

336807-500MG 336807-2G
220057-062EA 310.77 USD
220057-062 220057-919
Iridium(III) chloride
Iridium(III) chloride
Formula: Cl₃Ir
MW: 298.58 g/mol
Density: 5.3 g/cm³ (20 °C)
MDL Number: MFCD00011063
CAS Number: 10025-83-9
EINECS: 233-044-6
UN: 3260
ADR: 8,II
Merck Index: 12,05101

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR