Niobium(V) chloride

336602-50G 336602-10G
220057-900EA 576.92 USD
220057-900 220057-899
Niobium(V) chloride
Niobium(V) chloride
Formula: Cl₅Nb
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 205 °C
Density: 2.75 g/cm³ (20 °C)
MDL Number: MFCD00011127
CAS Number: 10026-12-7
EINECS: 233-059-8
UN: 3260
ADR: 8,III
Merck Index: 12,06649

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR