2-Amino-m-cresol ≥96%

Synonyms: 2-Hydroxy-6-methylaniline, 2-Amino-3-hydroxytoluene, 2-Amino-3-methylphenol, 1-Amino-2-hydroxy-6-methylbenzene

336572-5G 336572-25G
220057-894EA 102.85 USD
220057-894 220057-893
2-Amino-m-cresol ≥96%
2-Amino-m-cresol
Formula: C₇H₉NO
MW: 123.15 g/mol
MDL Number: MFCD00075082
CAS Number: 2835-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR