Lead(II) carbonate ACS

Danger

Synonyms: Lead carbonat

336378-100G 336378-500G
220057-865EA 52.98 USD
220057-865 220057-866
Lead(II) carbonate ACS
Lead(II) carbonate
Formula: PbCO₃
MW: 267.21 g/mol
Melting Pt: 315 °C
Density: 6.43 g/cm³ (20 °C)
MDL Number: MFCD00064631
CAS Number: 598-63-0
EINECS: 209-943-4
UN: 2291
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR